Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-m-Nitrobiphenyline oxalate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
945618-97-3
C19H19N3O7
401.375
QEAVQZOJQONCSF-UTONKHPSSA-N
r-+-m-nitrobiphenyline oxalate
16757280
2-[(1R)-1-[2-(3-nitrophenyl)phenoxy]ethyl]-4,5-dihydro-1H-imidazole;oxalic acid
CC(C1=NCCN1)OC2=CC=CC=C2C3=CC(=CC=C3)[N+](=O)[O-].C(=O)(C(=O)O)O
This item is not returnable. View return policy
945618-97-3
C19H19N3O7
401.375
QEAVQZOJQONCSF-UTONKHPSSA-N
r-+-m-nitrobiphenyline oxalate
16757280
2-[(1R)-1-[2-(3-nitrophenyl)phenoxy]ethyl]-4,5-dihydro-1H-imidazole;oxalic acid
CC(C1=NCCN1)OC2=CC=CC=C2C3=CC(=CC=C3)[N+](=O)[O-].C(=O)(C(=O)O)O

Potent α2C agonist

Quantity 10mg
Assay Percent Range >99%
Formula Weight 401.38
Chemical Name or Material (R)-(+)-m-Nitrobiphenyline oxalate

Recommended Storage : Store at +4°C