Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-Mandelic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7022137
Click to view available options
Quantity:
25 g
100 g
611-71-2
C8H8O3
152.15
MFCD00064251
IWYDHOAUDWTVEP-SSDOTTSWSA-N
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
11914
CHEBI:17656
(2R)-2-hydroxy-2-phenylacetic acid
O[C@@H](C(O)=O)C1=CC=CC=C1
611-71-2
C8H8O3
152.15
MFCD00064251
IWYDHOAUDWTVEP-SSDOTTSWSA-N
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
11914
CHEBI:17656
(2R)-2-hydroxy-2-phenylacetic acid
O[C@@H](C(O)=O)C1=CC=CC=C1

(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

Solubility
It is partly soluble in water, freely soluble in isopropyl and ethyl alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under ordinary conditions. Incompatabile to strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 130°C to 134°C
Solubility Information It is partly soluble in water,freely soluble in isopropyl and ethyl alcohol.
Formula Weight 152.15
Percent Purity 98%
Quantity 25 g
Optical Rotation -153° (c=5 in Water)
Sensitivity Light sensitive
Chemical Name or Material (R)-(-)-Mandelic acid
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 210-276-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit