Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-α-Methylleucine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
29589-03-5
C7H15NO2
145.2
ARSWQPLPYROOBG-SSDOTTSWSA-N
d-leucine, 2-methyl,r-2-amino-2,4-dimethylpentanoic acid,h-alpha-me-d-leu-oh,2r-2-amino-2,4-dimethylpentanoic acid,r-alpha-methylleucine,alpha-methyl-d-leucine,ambotzhaa5280,h-me-d-leu-oh,h-a-me-d-leu-oh,2-methyl-d-leucine
6951132
(2R)-2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
29589-03-5
C7H15NO2
145.2
ARSWQPLPYROOBG-SSDOTTSWSA-N
d-leucine, 2-methyl,r-2-amino-2,4-dimethylpentanoic acid,h-alpha-me-d-leu-oh,2r-2-amino-2,4-dimethylpentanoic acid,r-alpha-methylleucine,alpha-methyl-d-leucine,ambotzhaa5280,h-me-d-leu-oh,h-a-me-d-leu-oh,2-methyl-d-leucine
6951132
(2R)-2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
Specific Rotation -35
Melting Point >237°C
Color White to Yellow
Quantity 100mg
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5%
Formula Weight 145.2
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-α-Methylleucine, 98% ee
Show More Show Less