Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-α-Methyltyrosine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
672-86-6
C10H13NO3
195.22
NHTGHBARYWONDQ-SNVBAGLBSA-N
r-alpha-methyltyrosine,alpha-methyl-d-tyr,r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid,2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid,alpha-methyl-d-tyrosine,metirosinum inn-latin,metirosina inn-spanish,l-tyrosine, alpha-methyl-,-,d +-metyrosine,metyrosine usan
12650
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
672-86-6
C10H13NO3
195.22
NHTGHBARYWONDQ-SNVBAGLBSA-N
r-alpha-methyltyrosine,alpha-methyl-d-tyr,r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid,2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid,alpha-methyl-d-tyrosine,metirosinum inn-latin,metirosina inn-spanish,l-tyrosine, alpha-methyl-,-,d +-metyrosine,metyrosine usan
12650
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
Specific Rotation -18
Melting Point >211°C
Color White to Yellow
Infrared Spectrum Authentic
Enantiomeric Excess 98.0% min.
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 195.22
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-α-Methyltyrosine, 98% ee
Show More Show Less