Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
80548-31-8
C10H15NO
165.23
MFCD01862171
GXIWMXAAPLZOBY-SECBINFHSA-N
r-+-2-1-phenylethyl amino ethanol,2-1r-1-phenylethyl amino ethan-1-ol,2-1r-1-phenylethyl amino ethanol,2-r-alpha-methylbenzylamino ethanol,ethanol, 2-1r-1-phenylethyl amino,r-+-n-2-hydroxyethyl-phenylethylamine,r-+-n-2-hydroxyethyl-alpha-methylbenzylamine,r-+-n-2-hydroxyethyl-alpha-phenylethylamine
6993821
2-[[(1R)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
80548-31-8
C10H15NO
165.23
MFCD01862171
GXIWMXAAPLZOBY-SECBINFHSA-N
r-+-2-1-phenylethyl amino ethanol,2-1r-1-phenylethyl amino ethan-1-ol,2-1r-1-phenylethyl amino ethanol,2-r-alpha-methylbenzylamino ethanol,ethanol, 2-1r-1-phenylethyl amino,r-+-n-2-hydroxyethyl-phenylethylamine,r-+-n-2-hydroxyethyl-alpha-methylbenzylamine,r-+-n-2-hydroxyethyl-alpha-phenylethylamine
6993821
2-[[(1R)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
Infrared Spectrum Authentic
Specific Rotation Condition +44° (22°C c=1,C2H5OH)
Specific Rotation + 44.00
Quantity 10g
Color Colorless
Refractive Index 1.5320 to 1.5350
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 165.23
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (R)-(+)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA