Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N-Allyl-1-phenylethylamine hydrochloride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
204587-86-0
C11H15N·HCl
197.71
MFCD01862108
BAFILFKLBRDSLS-HNCPQSOCSA-N
r--n-allyl-1-phenylethylamine hydrochloride,1r-1-phenylethyl prop-2-en-1-yl amine hydrochloride,r-+-n-2-allyl phenylethylamine,r-n-1-phenylethyl prop-2-en-1-amine hydrochloride
53393158
N-[(1R)-1-phenylethyl]prop-2-en-1-amine;hydrochloride
CC(C1=CC=CC=C1)NCC=C.Cl
204587-86-0
C11H15N·HCl
197.71
MFCD01862108
BAFILFKLBRDSLS-HNCPQSOCSA-N
r--n-allyl-1-phenylethylamine hydrochloride,1r-1-phenylethyl prop-2-en-1-yl amine hydrochloride,r-+-n-2-allyl phenylethylamine,r-n-1-phenylethyl prop-2-en-1-amine hydrochloride
53393158
N-[(1R)-1-phenylethyl]prop-2-en-1-amine;hydrochloride
CC(C1=CC=CC=C1)NCC=C.Cl
Color White
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H5CH(CH3)NHCH2CHCH2·HCl
Formula Weight 197.71
Physical Form Powder and Chunks
Percent Purity 95%
Chemical Name or Material (R)-(+)-N-Allyl-1-phenylethylamine hydrochloride
Show More Show Less