Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-N-BOC-2-(5-Bromothienyl)alanine, 95%, 98% ee, Acros Organics
Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
Specifications
Melting Point | 68.8°C |
Quantity | 1g |
Packaging | Glass bottle |
Assay Percent Range | 94% min. (HPLC) |
Formula Weight | 350.23 |
Percent Purity | 95%,98% ee |
Chemical Name or Material | (R)-N-BOC-2-(5-Bromothienyl)alanine |