Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-BOC-2-(5-Bromothienyl)alanine, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
261380-16-9
C12H16BrNO4S
350.23
HHNNFRCLMQFXBA-MRVPVSSYSA-N
boc-d-2-5-bromothienyl alanine,r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoic acid,2r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-2-5-br-thiophenealanine,r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoicacid
7009118
(2R)-3-(5-bromothiophen-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(S1)Br)C(=O)O
261380-16-9
C12H16BrNO4S
350.23
HHNNFRCLMQFXBA-MRVPVSSYSA-N
boc-d-2-5-bromothienyl alanine,r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoic acid,2r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-2-5-br-thiophenealanine,r-3-5-bromothiophen-2-yl-2-tert-butoxycarbonyl amino propanoicacid
7009118
(2R)-3-(5-bromothiophen-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(S1)Br)C(=O)O
Melting Point 68.8°C
Quantity 500mg
Packaging Glass bottle
Assay Percent Range 94% min. (HPLC)
Formula Weight 350.23
Percent Purity 95%,98% ee
Chemical Name or Material (R)-N-BOC-2-(5-Bromothienyl)alanine