Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-BOC-2-Furylalanine 95%, (98% e.e.), ACROS Organics™

Click to view available options
Quantity:
1g
261380-18-1
C12H17NO5
255.27
YWLUWSMJXXBOLV-SECBINFHSA-N
r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid,r-n-boc-2-furylalanine,boc-d-3-2-furyl alanine,boc-d-3-2-furyl-alanine,2r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid,n-boc-d-2-furylalanine,2r-2-tert-butoxy carbonylamino-3-2-furyl propanoic acid,r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoicacid,2r-3-furan-2-yl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
2734480
(2R)-3-(furan-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CO1)C(=O)O
261380-18-1
C12H17NO5
255.27
YWLUWSMJXXBOLV-SECBINFHSA-N
r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid,r-n-boc-2-furylalanine,boc-d-3-2-furyl alanine,boc-d-3-2-furyl-alanine,2r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid,n-boc-d-2-furylalanine,2r-2-tert-butoxy carbonylamino-3-2-furyl propanoic acid,r-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoicacid,2r-3-furan-2-yl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
2734480
(2R)-3-(furan-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CO1)C(=O)O
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Color Brown-Orange or Orange
CAS Min % 94.0
CAS Max % 100.0
Formula Weight 255.27
Physical Form Viscous Liquid or Waxy Mass
Percent Purity ≥94%
Chemical Name or Material (R)-N-BOC-2-Furylalanine, 95%, (98% e.e.)