Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
250mg
500mg
1g
Packaging:
Glass bottle
161024-80-2
C14H19NO4
265.31
MFCD01320859
JTNQFJPZRTURSI-LLVKDONJSA-N
r-n-boc-3-amino-3-phenylpropanoic acid,r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,boc-r-3-amino-3-phenylpropionic acid,boc-beta-phe-oh,r-3-boc-amino-3-phenylpropionic acid,r-3-boc-amino-3-phenylpropanoic acid,boc-r-phenylalanine,3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid
7009108
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
161024-80-2
C14H19NO4
265.31
MFCD01320859
JTNQFJPZRTURSI-LLVKDONJSA-N
r-n-boc-3-amino-3-phenylpropanoic acid,r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,boc-r-3-amino-3-phenylpropionic acid,boc-beta-phe-oh,r-3-boc-amino-3-phenylpropionic acid,r-3-boc-amino-3-phenylpropanoic acid,boc-r-phenylalanine,3r-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,r-3-tert-butoxycarbonylamino-3-phenyl-propionic acid
7009108
(3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
Melting Point 123.4°C
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Quantity 250mg
Infrared Spectrum Authentic
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 265.31
Physical Form Powder
Percent Purity 95%
Chemical Name or Material (R)-N-BOC-3-Amino-3-phenylpropanoic acid, 98% ee
Show More Show Less