Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(R)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, 98% ee, Acros Organics
Click to view available options
Quantity:
250mg
500mg
1g
Packaging:
Glass bottle
Specifications
Enantiomeric Excess | 97.5% min. |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Color | White |
Quantity | 500mg |
Melting Point | 123.4°C |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Formula Weight | 265.31 |
Show More |