Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-BOC-piperidine-3-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
163438-09-3
C11H19NO4
229.28
NXILIHONWRXHFA-MRVPVSSYSA-N
r-boc-nipecotic acid,r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid,r-n-boc-piperidine-3-carboxylic acid,boc-d-nip-oh,r-1-boc-piperidine-3-carboxylic acid,r-1-boc-nipecotic acid,d-1-boc-nipecotic acid,n-boc-r-nipecotic acid,r-boc-nip,r-boc-nip-oh
643516
(3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
163438-09-3
C11H19NO4
229.28
NXILIHONWRXHFA-MRVPVSSYSA-N
r-boc-nipecotic acid,r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid,r-n-boc-piperidine-3-carboxylic acid,boc-d-nip-oh,r-1-boc-piperidine-3-carboxylic acid,r-1-boc-nipecotic acid,d-1-boc-nipecotic acid,n-boc-r-nipecotic acid,r-boc-nip,r-boc-nip-oh
643516
(3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
Melting Point 166°C to 171°C
Quantity 5g
Specific Rotation -52.2
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Specific Rotation Condition −52.20° (20°C c=1.15 MeOH)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 229.28
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material (R)-N-BOC-Piperidine-3-carboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning