Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (R)-N-Diphenylphosphino-N-methyl-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™

Asymmetric hydrogenation

Manufacturer:  Alfa Aesar™ 44684-77

Quantity 0.1g

 View more versions of this product

Catalog No. AA4468477

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

406680-94-2
611.5
(R)-Methyl BoPhoz™
C37H35FeNP2
MFCD05865220
(R)-N-Diphenylphosphino-N-methyl-[ (S)-2- (diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™

Specifications

406680-94-2
C37H35FeNP2
MFCD05865220
112° to 115°C
0.1g
−297° (C=1 in dichloromethane)
Solid
611.5
Yellow to orange
(R)-Methyl BoPhoz™
Odorless
(R)-N-Diphenylphosphino-N-methyl-[ (S)-2- (diphenylphosphino)ferrocenyl]ethylamine, (R)-Methyl BoPhoz™
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit