Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-2-Thienylalanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
201532-42-5
C22H19NO4S
393.457
MFCD00065677
PXBMQFMUHRNKTG-HXUWFJFHSA-N
fmoc-d-2-thienylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-3-2-thienyl-d-alanine,fmoc-d-3-2-thienyl-l-alanine,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-2-yl propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-d-thi-oh,fmoc-2-thienyl-d-ala-oh,fmoc-beta-2-thienyl-d-alanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-thiophen-2-ylpropanoic acid
2734454
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CS4)C(=O)O
201532-42-5
C22H19NO4S
393.457
MFCD00065677
PXBMQFMUHRNKTG-HXUWFJFHSA-N
fmoc-d-2-thienylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-3-2-thienyl-d-alanine,fmoc-d-3-2-thienyl-l-alanine,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-2-yl propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-d-thi-oh,fmoc-2-thienyl-d-ala-oh,fmoc-beta-2-thienyl-d-alanine,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-thiophen-2-ylpropanoic acid
2734454
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CS4)C(=O)O
Quantity 1g
Color White
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 393.46
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (R)-N-FMOC-2-Thienylalanine, 95%, (98% e.e.)
Show More Show Less