Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-3-Amino-3-phenylpropanoic acid, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
220498-02-2
C24H21NO4
387.435
MFCD01311783
PTSLRPMRTOVHAB-JOCHJYFZSA-N
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-r-3-amino-3-phenylpropionic acid,fmoc-l-beta-phenylalanine,fmoc-beta-phe-oh,r-n-fmoc-3-amino-3-phenylpropanoic acid,r-3-fmoc-amino-3-phenylpropionic acid,fmoc-r-phenylalanine,3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid,3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid,fmoc-d-?-phe-oh
2734302
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
220498-02-2
C24H21NO4
387.435
MFCD01311783
PTSLRPMRTOVHAB-JOCHJYFZSA-N
r-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-r-3-amino-3-phenylpropionic acid,fmoc-l-beta-phenylalanine,fmoc-beta-phe-oh,r-n-fmoc-3-amino-3-phenylpropanoic acid,r-3-fmoc-amino-3-phenylpropionic acid,fmoc-r-phenylalanine,3r-3-9h-fluoren-9-ylmethoxycarbonylamino-3-phenylpropanoic acid,3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid,fmoc-d-?-phe-oh
2734302
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Color White
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min.
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 387.43
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (R)-N-FMOC-3-Amino-3-phenylpropanoic acid, 95%, (98% e.e.)
Show More Show Less