Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-3-Thienylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
220497-90-5
C22H19NO4S
393.457
MFCD00151915
LSBZJMRHROCYGY-HXUWFJFHSA-N
fmoc-d-3-thienylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-beta-3-thienyl-d-ala-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-d-3-3-thienyl alanine,fmoc-d-3-thi-oh
6958726
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O
220497-90-5
C22H19NO4S
393.457
MFCD00151915
LSBZJMRHROCYGY-HXUWFJFHSA-N
fmoc-d-3-thienylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-beta-3-thienyl-d-ala-oh,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-d-3-3-thienyl alanine,fmoc-d-3-thi-oh
6958726
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Color White
Quantity 500mg
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 393.46
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (R)-N-FMOC-3-Thienylalanine, 95%, (98% e.e.)
Show More Show Less