Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-4-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
205526-34-7
C25H20N2O4
412.445
MFCD00672560
JOPKKUTWCGYCDA-HSZRJFAPSA-N
fmoc-4-cyano-d-phenylalanine,fmoc-d-phe 4-cn-oh,fmoc-d-4-cyanophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-cyanophenyl propanoic acid,fmoc-d-4-cyanophenylalanine,n-fmoc-d-4-cyanophenylalanine,2r-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2r-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-dphe pcn-oh,pubchem20601
2734467
(2R)-3-(4-cyanophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C#N)C(=O)O
205526-34-7
C25H20N2O4
412.445
MFCD00672560
JOPKKUTWCGYCDA-HSZRJFAPSA-N
fmoc-4-cyano-d-phenylalanine,fmoc-d-phe 4-cn-oh,fmoc-d-4-cyanophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-cyanophenyl propanoic acid,fmoc-d-4-cyanophenylalanine,n-fmoc-d-4-cyanophenylalanine,2r-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2r-3-4-cyanophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-dphe pcn-oh,pubchem20601
2734467
(2R)-3-(4-cyanophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)C#N)C(=O)O
Quantity 1g
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 412.44
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (R)-N-FMOC-4-Cyanophenylalanine, 95%, (98% e.e.)
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA