Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
193086-74-7
C25H22FNO4
419.46
VNYGHKCAXQKZEQ-VWLOTQADSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid,s-n-fmoc-alpha-methyl-2-fluorophenylalaine,fmoc-alpha-methyl-d-2-fluorophe,fmoc-alpha-methyl-d-2-fluorophenylalanine,r-n-fmoc-a-methyl-2-fluorophenylalanine
44598473
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
193086-74-7
C25H22FNO4
419.46
VNYGHKCAXQKZEQ-VWLOTQADSA-N
s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid,s-n-fmoc-alpha-methyl-2-fluorophenylalaine,fmoc-alpha-methyl-d-2-fluorophe,fmoc-alpha-methyl-d-2-fluorophenylalanine,r-n-fmoc-a-methyl-2-fluorophenylalanine
44598473
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
CC(CC1=CC=CC=C1F)(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Specific Rotation 48
Infrared Spectrum Authentic
Quantity 500mg
Melting Point 111.0°C to 122.0°C
Packaging Glass bottle
Enantiomeric Excess 98.0% min.
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 419.46
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98% ee
Show More Show Less