Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
616867-28-8,1634-04-4
C21H23NO4
353.43
AWEZXIRZNQCCNN-OAQYLSRUSA-N
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid,l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl,r-n-fmoc-alpha-methylvaline,fmoc-r-2-amino-2,3-dimethylbutanoicacid,fmoc-r-2-amino-2,3-dimethylbutanoic acid,n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline,r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid,r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
11210384
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
616867-28-8,1634-04-4
C21H23NO4
353.43
AWEZXIRZNQCCNN-OAQYLSRUSA-N
r-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid,l-isovaline, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl,r-n-fmoc-alpha-methylvaline,fmoc-r-2-amino-2,3-dimethylbutanoicacid,fmoc-r-2-amino-2,3-dimethylbutanoic acid,n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-l-isovaline,r-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid,r-2-9h-fluorene-9-ylmethoxycarbonylamino-2,3-dimethylbutyric acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid
11210384
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Melting Point 37°C to 53°C
Specific Rotation -8.6
Enantiomeric Excess 98.0% min.
Residual Solvents 10% max. (MTBE)
Color White to Yellow
Infrared Spectrum Authentic
Quantity 100mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 353.43
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-N-FMOC-α-Methylvaline, 98% ee
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning