Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-FMOC-Styrylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
215190-23-1
C26H23NO4
413.473
MFCD01311775
ZFMHHKMOLFNMMV-NOENIZQJSA-N
fmoc-d-styrylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-phenylpent-4-enoic acid,fmoc-3-styryl-d-alanine,fmoc-beta-styryl-d-ala-oh,fmoc-d-ala styryl-oh,fmoc-,a-styryl-d-ala-oh,r-2-fmoc-amino-5-phenyl-4-pentenoicacid,s-2-fmoc-amino-5-phenyl-4-pentenoic acid,e,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-5-phenylpent-4-enoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-phenyl-4-pentenoic acid
5702631
(E,2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-phenylpent-4-enoic acid
C1=CC=C(C=C1)C=CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
215190-23-1
C26H23NO4
413.473
MFCD01311775
ZFMHHKMOLFNMMV-NOENIZQJSA-N
fmoc-d-styrylalanine,r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-phenylpent-4-enoic acid,fmoc-3-styryl-d-alanine,fmoc-beta-styryl-d-ala-oh,fmoc-d-ala styryl-oh,fmoc-,a-styryl-d-ala-oh,r-2-fmoc-amino-5-phenyl-4-pentenoicacid,s-2-fmoc-amino-5-phenyl-4-pentenoic acid,e,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-5-phenylpent-4-enoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-phenyl-4-pentenoic acid
5702631
(E,2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-phenylpent-4-enoic acid
C1=CC=C(C=C1)C=CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Quantity 1g
Enantiomeric Excess 97.5% min.
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 413.47
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (R)-N-FMOC-Styrylalanine, 95%, (98% e.e.)
Show More Show Less