Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-α-Propargylglycine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
23235-03-2
C5H7NO2
113.12
DGYHPLMPMRKMPD-SCSAIBSYSA-N
d-propargylglycine,r-2-aminopent-4-ynoic acid,r-alpha-propargylglycine,unii-2252qef2pl,4-pentynoic acid, 2-amino-, 2r,2r-2-aminopent-4-ynoic acid,r-2-propargylglycine,r-2-amino-4-pentynoic acid,d-2-propynylglycine,propargylglycine, d
6992713
(2R)-2-aminopent-4-ynoic acid
C#CCC(C(=O)O)N
23235-03-2
C5H7NO2
113.12
DGYHPLMPMRKMPD-SCSAIBSYSA-N
d-propargylglycine,r-2-aminopent-4-ynoic acid,r-alpha-propargylglycine,unii-2252qef2pl,4-pentynoic acid, 2-amino-, 2r,2r-2-aminopent-4-ynoic acid,r-2-propargylglycine,r-2-amino-4-pentynoic acid,d-2-propynylglycine,propargylglycine, d
6992713
(2R)-2-aminopent-4-ynoic acid
C#CCC(C(=O)O)N
Melting Point >180°C
Specific Rotation 31.65
Packaging Glass Bottle
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Color White to Yellow
Quantity 500mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5%
Formula Weight 113.12
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (R)-α-Propargylglycine, 98% ee
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning