Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-(-)-1,2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine]rhodium(I) BF4, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
56977-92-5
C36H40BF4O2P2Rh-
756.371
MFCD01074456
DCGUTTVLWMBBAX-BSLNZZRFSA-N
(R,R)-(-)-1,2-Bis[(o-methoxyphenyl)phenylphosphino]ethane(1,5-cyclooctadiene)rhodium(I)terafluoroborate
132274297
(5Z)-cycloocta-1,5-diene;(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.COC1=CC=CC=C1P(CCP(C2=CC=CC=C2)C3=CC=CC=C3OC)C4=CC=CC=C4.C1CC=CCCC=C1.[Rh]
56977-92-5
C36H40BF4O2P2Rh-
756.371
MFCD01074456
DCGUTTVLWMBBAX-BSLNZZRFSA-N
(R,R)-(-)-1,2-Bis[(o-methoxyphenyl)phenylphosphino]ethane(1,5-cyclooctadiene)rhodium(I)terafluoroborate
132274297
(5Z)-cycloocta-1,5-diene;(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.COC1=CC=CC=C1P(CCP(C2=CC=CC=C2)C3=CC=CC=C3OC)C4=CC=CC=C4.C1CC=CCCC=C1.[Rh]
Quantity 100mg
Packaging Glass bottle
Color Orange
Infrared Spectrum Authentic
Formula Weight 756.38
Physical Form Crystalline Powder
Chemical Name or Material (R, R)-(-)-1, 2-Ethanediylbis[(2-methoxyphenyl)phenylphosphine]rhodium(I) BF4