Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-Rolipram, Tocris Bioscience™

Catalog Number p-4783098
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
85416-75-7
C16H21NO3
275.348
HJORMJIFDVBMOB-LBPRGKRZSA-N
r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
448055
CHEBI:40133
(4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one
COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3
This item is not returnable. View return policy
85416-75-7
C16H21NO3
275.348
HJORMJIFDVBMOB-LBPRGKRZSA-N
r---rolipram, r-rolipram, r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, --rolipram, 4r-4-3-cyclopentyloxy-4-methoxyphenyl pyrrolidin-2-one, unii-dpx51kup08, dpx51kup08, 1xmy, rolipram, r
448055
CHEBI:40133
(4R)-4-(3-cyclopentyloxy-4-methoxyphenyl)pyrrolidin-2-one
COC1=C(C=C(C=C1)C2CC(=O)NC2)OC3CCCC3

PDE4 inhibitor. More active enantiomer of rolipram (Cat. No. 0905)

More active enantiomer of the PDE4 inhibitor rolipram; 2-10-fold more potent than the S-(+) enantiomer.

Racemate and S-enantiomer also available.

Quantity 10 mg
Formula Weight 275.35
Percent Purity >98%
Chemical Name or Material (R)-(-)-Rolipram
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit