Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cayman ChemicalSupplier Diversity Partner (R,S)-Anatabine, 2743-90-0, 50 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 1100150MG
A minor alkaloid produced in plants of the Solanaceae family, including tobacco; detection in urine is used as an indicator of tobacco use; diminishes Aβ production, reduces the transcription and protein levels of β-secretase, and dose dependently inhibits NF-κB activation
Catalog No. NC1468737