Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-tert-Butylsulfinamide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
196929-78-9
C4H11NOS
121.2
MFCD05861479
CESUXLKAADQNTB-SSDOTTSWSA-N
r-+-2-methyl-2-propanesulfinamide,r-2-methylpropane-2-sulfinamide,r-+-t-butylsulfinamide,r-+-tert-butanesulfinamide,r-+-tert-butylsulfinamide,r-2-methyl-2-propanesulfinamide,r-tert-butanesulfinamide,r-+-tert-butyl sulphinamide,r-tert-butylsulfinamide,r-+-tert-butyl sulfinamide
10964479
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
196929-78-9
C4H11NOS
121.2
MFCD05861479
CESUXLKAADQNTB-SSDOTTSWSA-N
r-+-2-methyl-2-propanesulfinamide,r-2-methylpropane-2-sulfinamide,r-+-t-butylsulfinamide,r-+-tert-butanesulfinamide,r-+-tert-butylsulfinamide,r-2-methyl-2-propanesulfinamide,r-tert-butanesulfinamide,r-+-tert-butyl sulphinamide,r-tert-butylsulfinamide,r-+-tert-butyl sulfinamide
10964479
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
Specific Rotation 3.0° to 5.0° (20°C, 589nm) (c=1, chloroform)
Infrared Spectrum Authentic
Quantity 1g
Color Undesignated
Melting Point 99.0°C to 102.0°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 % min. (GC)
Linear Formula (CH3)3CSONH2
Merck Index 15,3607
Formula Weight 121.2
Physical Form Powder
Percent Purity 98%
Chemical Name or Material (R)-(+)-tert-Butylsulfinamide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.