Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Tetrahydro-2-furoic acid, 99% (98% ee), ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
87392-05-0
C5H8O3
116.116
MFCD00211271
UJJLJRQIPMGXEZ-SCSAIBSYSA-N
r-+-2-tetrahydrofuroic acid,r-+-tetrahydro-2-furoic acid,r-tetrahydrofuran-2-carboxylic acid,r-+-tetrahydrofuran-2-carboxylic acid,r-2-tetrahydrofuroic acid,2-furancarboxylic acid, tetrahydro-, 2r,2r-oxolane-2-carboxylic acid,unii-7396hfq44j,r-tetrahydro-2-furoic acid,d-tetrahydro-furan-2-carboxylic acid
2734707
(2R)-oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O
87392-05-0
C5H8O3
116.116
MFCD00211271
UJJLJRQIPMGXEZ-SCSAIBSYSA-N
r-+-2-tetrahydrofuroic acid,r-+-tetrahydro-2-furoic acid,r-tetrahydrofuran-2-carboxylic acid,r-+-tetrahydrofuran-2-carboxylic acid,r-2-tetrahydrofuroic acid,2-furancarboxylic acid, tetrahydro-, 2r,2r-oxolane-2-carboxylic acid,unii-7396hfq44j,r-tetrahydro-2-furoic acid,d-tetrahydro-furan-2-carboxylic acid
2734707
(2R)-oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O
Density 1.2100g/mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 1g
Specific Rotation +18° to +20° (20°C, 589nm) (neat)
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Formula Weight 116.12
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material (R)-(+)-Tetrahydro-2-furoic acid, 99% (98% e.e.)
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger