Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-Tetrahydrofurfurylamine, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7202-43-9
C5H11NO
101.15
MFCD00192476
YNOGYQAEJGADFJ-RXMQYKEDSA-N
r---tetrahydrofurfurylamine,r-tetrahydrofuran-2-yl methanamine,r-tetrahydrofurfurylamine,2r-oxolan-2-ylmethanamine,2r-tetrahydro-2-furanmethanamine,2-furanmethanamine, tetrahydro-, 2r,1-2r-tetrahydrofuran-2-yl methanamine,pubchem7001,2r-tetrahydrofurfurylamine
2734124
[(2R)-oxolan-2-yl]methanamine
C1CC(OC1)CN
7202-43-9
C5H11NO
101.15
MFCD00192476
YNOGYQAEJGADFJ-RXMQYKEDSA-N
r---tetrahydrofurfurylamine,r-tetrahydrofuran-2-yl methanamine,r-tetrahydrofurfurylamine,2r-oxolan-2-ylmethanamine,2r-tetrahydro-2-furanmethanamine,2-furanmethanamine, tetrahydro-, 2r,1-2r-tetrahydrofuran-2-yl methanamine,pubchem7001,2r-tetrahydrofurfurylamine
2734124
[(2R)-oxolan-2-yl]methanamine
C1CC(OC1)CN
Boiling Point 151°C to 152°C
Flash Point 46°C
Specific Gravity 0.98
Specific Rotation -13.5
Refractive Index 1.4540 to 1.456
Specific Rotation Condition − 13.50 (20.00°C c−4,MeOH)
Quantity 1g
Density 0.9800g/mL
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Solubility Information Solubility in water: miscible with water
Formula Weight 101.15
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Warning