Learn More
Alfa Aesar™ R15K, HIV-1 Inhibitory Peptide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66327EXD
 View more versions of this product
Catalog No. AAJ66327EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1655.97
0.5mg
Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys
-30° to -10°C
HPLC
Solid
RUO
R15K, HIV-1 Inhibitory Peptide