Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ R4410 Series TLS 2200™ and TLS-PC Link™ Printer Ribbons

Developed to meet performance requirements of TLS2200 and TLS PC Link materials to be printed. Brady™ R4410 Series TLS 2200™ and TLS-PC Link™ Printer Ribbons are avialable in a variety of colors.

Supplier:  Brady™ R4410RD

 View more versions of this product

Catalog No. 19-104-659


Add to Cart

Description

Description

  • Ribbon size is 2 in. x 75 ft. (50.80 mm x 22.4 m)
Specifications

Specifications

Printer Ribbon
TLS 2200 Portable Thermal Transfer Printer, Brady TLS-PC Link Thermal Labeling System
2 in.
50.80 mm
Cartridge
Red
75 ft.
22.90 m
Ribbon Series: R4410
1 Ea.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit