Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ R6700 Series Thermal Transfer Printer Ribbons

Offers high performance against scratching, smudging, and harsh chemicals. Brady R6700 Series Thermal Transfer Printer Ribbons is made of premium wax/resin and can be used for printing on Brady's Black PermaSleeve materials.

$184.53 - $298.20

Specifications

Product Type R6700 series thermal transfer printer ribbon
Material Resin, Wax
Color White
Format Open-core Ribbon
Length (Metric) 299.9 m
View More Specs

Products 3
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Width (English) Price Quantity & Availability  
19-104-652
View Documents
Brady™
R6702WT
8.3 cm 3.27 in.
Each for $244.26
Add to cart
 
19-104-651
View Documents
Brady™
R6700WT
5.9 cm 2.36 in.
Each for $184.53
Add to cart
 
19-104-653
View Documents
Brady™
R6707WT
10.9 cm 4.33 in.
Each for $298.20
Add to cart
 
Description

Description

  • Offers high performance against scratching, smudging, and harsh chemicals
  • Used for printing on Brady Black PermaSleeve materials
Specifications

Specifications

R6700 series thermal transfer printer ribbon
White
299.9 m
25.4 mm I.D.
Resin, Wax
Open-core Ribbon
984 ft.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit