Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab11A Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001086
Description
Rab11A Polyclonal specifically detects Rab11A in Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
Rab11A | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Expected from sequence similarity: Human, Rat and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_004654 | |
RAB11A | |
Peptide with sequence TENKPKVQCCQNI corresponding to C-Terminus according to NP_004654.1. | |
0.1 mg | |
Cancer, Tumor Suppressors | |
8766 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction