Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$212.55 - $436.00
Specifications
Antigen | Rab11A |
---|---|
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Rab11A Polyclonal specifically detects Rab11A in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
Rab11A | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A | |
Recombinant protein of human Rab11A. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
8766 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title