Learn More
RAB11FIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181361AP
Catalog No. 50-173-2006
Add to cart
Description
Description
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
Specifications
Specifications
RAB11FIP2
0.79 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
G3XA57, Q7L804
KIAA0941, nRip11, Rab11 FIP2, RAB11FIP2
Rabbit
IgG
150 μL
-20°C
Primary
22841, 308003, 74998
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RAB11FIP2
Liquid
Rab11fip2
RAB11FIP2 Fusion Protein Ag12736
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat