Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB17 Mouse anti-Human, Clone: OTI4E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525126

Catalog No. PIMA525126


Add to cart

Description

Description

The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase (Lutcke et al., 1993 [PubMed 8486736]).
Specifications

Specifications

RAB17
Monoclonal
0.61 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H0T7
RAB17; RAB17, member RAS oncogene family ; AW413472
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64284
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4E7
Unconjugated
RAB17
Liquid
RAB17
Full length human recombit protein of RAB17 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit