Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab1A Mouse anti-Human, Mouse, Clone: 7H4, Novus Biologicals™

Mouse Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen Rab1A
Clone 7H4
Concentration 1 mg/ml
Dilution Western Blot 1:1000, Immunoprecipitation
Applications Western Blot, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB120204
View Documents
Novus Biologicals
NBP31104225UG
25 μg
Each for $156.50
Add to cart
 
NB120203
View Documents
Novus Biologicals
NBP311042100UG
100 μg
Each for $419.00
Add to cart
 
Description

Description

Rab1A Monoclonal antibody specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

Rab1A
1 mg/ml
Western Blot, Immunoprecipitation
Purified
RUO
PBS (pH 7.4), 50% Glycerol
5861
IgG1
Primary
Store at -20°C.
7H4
Western Blot 1:1000, Immunoprecipitation
Unconjugated
Mouse
Human, Mouse
DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A
Full length recombinant Rab1A protein (NP_004152.1)
Monoclonal
Protein G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit