Learn More
RAB1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178241AP
Catalog No. 50-173-1819
Add to cart
Description
Description
The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB1B regulates vesicular transport between the endoplasmic reticulum and successive Golgi compartments. Plays a role in the initial events of the autophagic vacuole development which take place at specialized regions of the endoplasmic reticulum.
Specifications
Specifications
RAB1B
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D1G1, Q9H0U4
RAB1B, Ras related protein Rab 1B
Rabbit
IgG
150 μL
-20°C
Primary
76308, 81876
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RAB1B
Liquid
Rab1b
RAB1B Fusion Protein Ag12197
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse