Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB1B Mouse anti-Human, Clone: 1673CT667.16.73, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524749

Catalog No. PIMA524749

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB1B regulates vesicular transport between the endoplasmic reticulum and successive Golgi compartments. Plays a role in the initial events of the autophagic vacuole development which take place at specialized regions of the endoplasmic reticulum.
Specifications

Specifications

RAB1B
Monoclonal
0.87 mg/mL
PBS with 0.09% sodium azide
Q9H0U4
Ras-related protein Rab-1B, RAB1B
Mouse
IgG1, kappa
200 μL
-20°C
Primary
81876
Western Blot
1673CT667.16.73
Unconjugated
RAB1B
Liquid
RAB1B
Recombit protein between 1-201 amino acids from human RAB1B
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit