Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB23 Monoclonal Antibody (OTI4C7), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527143
Description
RAB23 Monoclonal antibody specifically detects RAB23 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RAB23 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ULC3 | |
RAB23 | |
Human recombinant protein fragment corresponding to amino acids 1-234 of RAB23 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C7 | |
Unconjugated | |
RAB23 | |
AW545388; DKFZp781H0695; HSPC137; MGC8900; opb; opb2; open brain 2; OTTHUMP00000040021; protein open brain; RAB 23; RAB family small GTP binding protein RAB 23; rab-15; RAB23; RAB23, member RAS oncogene family; ras-related protein Rab-23; RP3-496N17.3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51715 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction