Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB24 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104070
Description
Antibody detects endogenous levels of total RAB24.
RAB24 may be involved in autophagy-related processes.
Specifications
RAB24 | |
Polyclonal | |
Unconjugated | |
RAB24 | |
6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19336, 361208, 53917 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35290, Q969Q5 | |
RAB24 | |
A synthesized peptide derived from human RAB24(Accession Q969Q5), corresponding to amino acid residues E126-A176. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction