Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB26 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | RAB26 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158882
|
Novus Biologicals
NBP158882 |
100 μL |
Each of 1 for $436.00
|
|
Description
RAB26 Polyclonal specifically detects RAB26 in Human, Mouse samples. It is validated for Western Blot.Specifications
RAB26 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Q9ULW5 | |
25837 | |
Synthetic peptides corresponding to RAB26(RAB26, member RAS oncogene family) The peptide sequence was selected from the N terminal of RAB26. Peptide sequence SRKKTPKSKGASTPAASTLPTANGARPARSGTALSGPDAPPNGPLQPGRP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
RAB26, member RAS oncogene family, Ras-related oncogene protein, ras-related protein Rab-26, V46133 | |
RAB26 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title