Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab27a Mouse anti-Human, Clone: 2.00E+02, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259459100UL

 View more versions of this product

Catalog No. NBP259459X


Add to cart

Description

Description

Rab27a Monoclonal antibody specifically detects Rab27a in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Rab27a
200
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A
Mouse
IgG1 Kappa
100 ul
Signal Transduction
Primary
5873
Western Blot, ELISA
1 mg/mL
Western Blot 1:3000, ELISA
Purified
RAB27A
Recombinant human RAB27A (1-221aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only