Learn More
Rab27a Mouse anti-Human, DyLight 755, Clone: 4D3F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237241IR
Catalog No. NBP237241IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Rab27a
4D3F11
Western Blot, ELISA
Purified
RAB27A
Purified recombinant fragment of human RAB27A (AA: FULL(1-221)) expressed in E. Coli.
Protein A or G purified
Signal Transduction
Primary
5873
ELISA, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human