Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab27a Mouse anti-Human, FITC, Clone: 4D3F11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237241F

Catalog No. NBP237241F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Rab27a
4D3F11
ELISA
Purified
RAB27A
Purified recombinant fragment of human RAB27A (AA: FULL(1-221)) expressed in E. Coli.
Protein A or G purified
Signal Transduction
Primary
5873
ELISA
FITC
PBS with 0.05% Sodium Azide
GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit