Learn More
Invitrogen™ RAB27A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110257
Description
Immunogen sequence: MSDGDYDYLI KFLALGDSGV GKTSVLYQYT DGKFNSKFIT TVGIDFREKR VVYRASGPDG ATGRGQRIHL QLWDTAGQER FRSLTTAFFR DAMGFLLLFD LTNEQSFLNV RNWISQLQMH AYCENPDIVL CGNKSDLEDQ RVVKEEEAIA LAEKYGIPYF ETSAANGTNI SQAIEMLLDL IMKRMERCVD KSWIPEGVVR SNGHASTDQL SEEKEKGACG C.
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
Specifications
RAB27A | |
Polyclonal | |
Unconjugated | |
RAB27A | |
2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; ashen; GS2; GTP-binding protein Ram; HsT18676; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; RAB27; rab-27; Rab27a; RAB27A, member RAS oncogene family; ram; Ram p25; ras-related protein Rab-27A | |
Rabbit | |
Affinity chromatography | |
RUO | |
11891, 5873 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.83 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51159, Q9ERI2 | |
RAB27A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human RAB27A (NP_899057.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.