Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB2A Mouse anti-Human, Clone:, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524761
Description
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.
Specifications
RAB2A | |
Monoclonal | |
ascites with 0.02% sodium azide | |
P61019 | |
RAB2A | |
human RAB2A recombit protein | |
RUO | |
5862 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
RAB2A | |
9330148M11Rik;C80220;GTP binding protein;LHX;OTTMUSP00000004796;RAB2;RAB2 member RAS oncogene family;RAB2A;RAB2A, member RAS oncogene family;RAB2A_HUMAN;Ras related protein Rab 2A;RAS-associated protein RAB2;Ras-related protein Rab-2A;RP23-183L6.1;Small GTP binding protein RAB2A. | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction