Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554389
Description
Immunogen sequence: GENVREFFFR VAALTFEANV LAELEKSGAR RIGDVVRINS DDSNLYLTAS KKKPTCCP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Protein transport. Involved in the redistribution of lysosomes to the peri-Golgi region. Plays a role in the maturation of phagosomes that engulf pathogens, such as S. aureus and M. tuberculosis. Plays a role in the fusion of phagosomes with lysosomes.
Specifications
RAB34 | |
Polyclonal | |
Unconjugated | |
RAB34 | |
AI326479; GTP-binding protein rah; Narr; Nine amino-acid residue-repeats; RAB34; RAB34, member of RAS oncogene family; RAB34, member RAS oncogene family; RAB39; rab-39; RAH; Rah1; Ras-related homolog; Ras-related protein RAB34; ras-related protein Rab-34; Ras-related protein Rab-34, isoform NARR; Ras-related protein Rab-39; ras-related protein Rah | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZG1 | |
RAB34 | |
Recombinant protein corresponding to Human RAB34. Recombinant protein control fragment (Product #RP-93224). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction