Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB37 Mouse anti-Human, Clone: OTI2B12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526105

Catalog No. PIMA526105


Add to cart

Description

Description

Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins, see MIM 179508.[supplied by OMIM, Apr 2006].
Specifications

Specifications

RAB37
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96AX2
RAB37
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
326624
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B12
Unconjugated
RAB37
Liquid
RAB37
Full length human recombit protein of RAB37 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit