Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB39 Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299624F
Description
RAB39 Polyclonal antibody specifically detects RAB39 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RAB39 | |
FITC | |
PBS | |
rab-39, RAB39, member RAS oncogene family, RAB39A, rab-related GTP-binding protein, ras-related protein Rab-39A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RAB39. | |
Antigen Affinity-purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Signal Transduction | |
Primary | |
54734 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title