Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB39B Antibody, mFluor Violet 500 SE, Novus Biologicals™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297201MFV500
Description
RAB39B Polyclonal antibody specifically detects RAB39B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
RAB39B | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
MRX72, RAB39B, member RAS oncogene family, ras-related protein Rab-39B | |
Produced in rabbits immunized with E. coli-derived Human RAB39B fragment. | |
0.1 mL | |
Signal Transduction | |
116442 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction