Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3IL1 Mouse anti-Human, Clone: OTI1B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526019

Catalog No. PIMA526019


Add to cart

Description

Description

Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.
Specifications

Specifications

RAB3IL1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TBN0
GRAB, guanine nucleotide exchange factor for Rab-3A, guanine nucleotide exchange factor for Rab3A, rab-3A-interacting-like protein 1, rab3A-interacting-like protein 1, rabin3-like 1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5866
Flow Cytometry, Western Blot
OTI1B5
Unconjugated
RAB3IL1
Liquid
RAB3IL1
Full length human recombit protein of RAB3IL1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit